Assay Details
| Gene Symbol: Il1rapl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160403.1 | NP_001153875.1 | 8 - 9 | 1697 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il1rapl1| Gene Symbol: Il1rapl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160403.1 | NP_001153875.1 | 8 - 9 | 1697 | 61 | |
Set Membership: |
|