Assay Details
| Gene Symbol: Ripk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138952.3 | NP_620402.1 | 1 - 2 | 276 | 132 | |
| GenBank mRNA | AF461040.1 | - | 1 - 2 | 174 | 132 | |
| AF487539.1 | - | 1 - 2 | 174 | 132 | ||
| AK159593.1 | - | 1 - 2 | 300 | 132 | ||
| AK172319.1 | - | 1 - 2 | 298 | 132 | ||
| AK172417.1 | - | 1 - 2 | 298 | 132 | ||
| BC050845.1 | - | 1 - 2 | 289 | 132 | ||
| BC058088.1 | - | 1 - 2 | 421 | 132 | ||
| BC069878.1 | - | 1 - 2 | 301 | 5316066 | 132 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ripk2
Genomic Map