Assay Details
| Gene Symbol: Runx1t1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111026.2 | NP_001104496.1 | 4 - 5 | 904 | 70 | |
| NM_001111027.2 | NP_001104497.1 | 5 - 6 | 1098 | 70 | ||
| NM_009822.3 | NP_033952.1 | 4 - 5 | 836 | 70 | ||
| GenBank mRNA | AB221624.1 | - | 3 - 4 | 483 | 70 | |
| AK032132.1 | - | 4 - 5 | 771 | 70 | ||
| AK142006.1 | - | 4 - 5 | 905 | 70 | ||
| BC139200.1 | - | 4 - 5 | 562 | 8862222 | 70 | |
| BC139201.1 | - | 4 - 5 | 566 | 8862223 | 70 | |
| D32007.1 | - | 4 - 5 | 836 | 70 | ||
| X79989.1 | - | 4 - 5 | 543 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Runx1t1
Genomic Map