Assay Details
| Gene Symbol: Lmna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002011.3 | NP_001002011.2 | 10 - 11 | 1956 | 92 | |
| GenBank mRNA | AK004619.2 | - | 10 - 11 | 1911 | 92 | |
| AK079972.1 | - | 2 - 3 | 117 | 92 | ||
| AK147150.1 | - | 10 - 11 | 1912 | 92 | ||
| AK153295.1 | - | 10 - 11 | 1853 | 92 | ||
| AK161221.1 | - | 10 - 11 | 1777 | 92 | ||
| BC015302.1 | - | 10 - 11 | 1832 | 4240057 | 92 | |
| BC094020.1 | - | 10 - 11 | 1818 | 4187374 | 92 | |
| D13181.1 | - | 7 - 8 | 1005 | 92 | ||
| DQ832702.1 | - | 10 - 11 | 1707 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lmna
Genomic Map