Assay Details
| Gene Symbol: Lmna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002011.3 | NP_001002011.2 | 11 - 12 | 2220 | 67 | |
| GenBank mRNA | AK004619.2 | - | 11 - 12 | 2175 | 67 | |
| AK079972.1 | - | 3 - 4 | 381 | 67 | ||
| AK147150.1 | - | 11 - 12 | 2176 | 67 | ||
| AK153295.1 | - | 11 - 12 | 2117 | 67 | ||
| AK161221.1 | - | 11 - 12 | 2041 | 67 | ||
| BC015302.1 | - | 11 - 12 | 2096 | 4240057 | 67 | |
| BC094020.1 | - | 11 - 12 | 2082 | 4187374 | 67 | |
| D13181.1 | - | 8 - 9 | 1269 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lmna
Genomic Map