Assay Details
| Gene Symbol: Spon2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133903.3 | NP_598664.3 | 2 - 3 | 512 | 92 | |
| GenBank mRNA | AK028987.1 | - | 2 - 3 | 457 | 92 | |
| AK042840.1 | - | 2 - 3 | 460 | 92 | ||
| AK131131.1 | - | 1 - 2 | 95 | 92 | ||
| AY457639.1 | - | 2 - 3 | 247 | 92 | ||
| BC017616.1 | - | 2 - 3 | 445 | 4923292 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Spon2
Genomic Map