Assay Details
Gene Symbol: Preb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001294302.1 | NP_001281231.1 | 4 - 5 | 852 | 102 | |
NM_016703.3 | NP_057912.2 | 4 - 5 | 852 | 102 | ||
GenBank mRNA | AF150808.1 | - | 4 - 5 | 755 | 102 | |
AF193017.1 | - | 4 - 5 | 764 | 102 | ||
AK013330.1 | - | 4 - 5 | 806 | 102 | ||
AK029838.1 | - | 4 - 5 | 103 | |||
AK036866.1 | - | 4 - 5 | 790 | 102 | ||
AK151288.1 | - | 4 - 5 | 834 | 102 | ||
AK188795.1 | - | 2 - 3 | 165 | 102 | ||
AK213634.1 | - | 2 - 3 | 167 | 102 | ||
BC017527.1 | - | 4 - 5 | 755 | 4921119 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
