Assay Details
| Gene Symbol: Ccr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009914.4 | NP_034044.3 | 1 - 2 | 201 | 98 | |
| GenBank mRNA | AK041106.1 | - | 1 - 2 | 202 | 98 | |
| AK089875.1 | - | 1 - 2 | 202 | 98 | ||
| AK089895.1 | - | 1 - 2 | 73 | 98 | ||
| AK153518.1 | - | 1 - 2 | 202 | 98 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK041106.1, AK089875.1, AK089895.1, AK153518.1, NM_009914.4 transcripts.
Set Membership: |
|

Ccr3
Genomic Map