Assay Details
Gene Symbol: Corin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122756.1 | NP_001116228.1 | 18 - 19 | 2469 | 115 | |
NM_016869.3 | NP_058565.2 | 18 - 19 | 2626 | 115 | ||
GenBank mRNA | AB013874.1 | - | 18 - 19 | 2626 | 115 | |
BC093485.1 | - | 18 - 19 | 2402 | 30648458 | 115 | |
BC138339.1 | - | 18 - 19 | 2609 | 8861359 | 115 | |
BC138340.1 | - | 18 - 19 | 2609 | 8861360 | 115 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
