Assay Details
| Gene Symbol: Slc4a1ap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009206.2 | NP_033232.2 | 5 - 6 | 1210 | 67 | |
| GenBank mRNA | AF035526.1 | - | 6 - 7 | 1093 | 67 | |
| AK138398.1 | - | 5 - 6 | 1165 | 67 | ||
| AK147161.1 | - | 5 - 6 | 548 | 67 | ||
| AK160250.1 | - | 4 - 5 | 1085 | 67 | ||
| BC072588.1 | - | 5 - 6 | 1193 | 67 | ||
| BC090964.1 | - | 5 - 6 | 762 | 30625511 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc4a1ap
Genomic Map