Assay Details
| Gene Symbol: Cad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289522.1 | NP_001276451.1 | 35 - 36 | 5762 | 58 | |
| NM_001289523.1 | NP_001276452.1 | 35 - 36 | 5762 | 58 | ||
| NM_023525.2 | NP_076014.1 | 35 - 36 | 5762 | 58 | ||
| GenBank mRNA | BC026620.1 | - | 5 - 6 | 691 | 58 | |
| BC043325.1 | - | 8 - 9 | 1291 | 58 | ||
| BC060119.1 | - | 35 - 36 | 5728 | 58 | ||
| BC060717.1 | - | 26 - 27 | 4219 | 58 | ||
| BC137856.1 | - | 35 - 36 | 5614 | 8860875 | 58 | |
| BC144972.1 | - | 35 - 36 | 5614 | 9053520 | 58 | |
| BC144973.1 | - | 35 - 36 | 5615 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cad
Genomic Map