Assay Details
| Gene Symbol: Nmu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019515.1 | NP_062388.1 | 8 - 9 | 591 | 113 | |
| GenBank mRNA | AF203444.1 | - | 8 - 9 | 591 | 113 | |
| AK077478.1 | - | 8 - 9 | 603 | 113 | ||
| BC138071.1 | - | 8 - 9 | 556 | 8861090 | 113 | |
More Information
Set Membership: |
|

Nmu
Genomic Map