Assay Details
| Gene Symbol: Lmbr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020295.3 | NP_064691.2 | 2 - 3 | 334 | 88 | |
| GenBank mRNA | AF190665.1 | - | 2 - 3 | 136 | 88 | |
| AK030631.1 | - | 3 - 4 | 1140 | 88 | ||
| BC016110.1 | - | 2 - 3 | 174 | 4481088 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lmbr1
Genomic Map