Assay Details
| Gene Symbol: Tmprss11g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177162.4 | NP_796136.2 | 4 - 5 | 410 | 131 | |
| GenBank mRNA | AK036981.1 | - | 4 - 5 | 411 | 131 | |
| BC118611.1 | - | 4 - 5 | 388 | 40107184 | 131 | |
| BC118660.1 | - | 4 - 5 | 387 | 40107187 | 131 | |
| CK627573.1 | - | 4 - 5 | 468 | 131 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmprss11g
Genomic Map