Assay Details
Gene Symbol: Ddx58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172689.3 | NP_766277.3 | 4 - 5 | 707 | 102 | |
GenBank mRNA | AK052871.1 | - | 4 - 5 | 692 | 102 | |
AK078287.1 | - | 4 - 5 | 639 | 102 | ||
AK128929.1 | - | 4 - 5 | 642 | 102 | ||
AK155930.1 | - | 4 - 5 | 643 | 102 | ||
AY553221.1 | - | 4 - 5 | 583 | 102 | ||
BC027369.1 | - | 4 - 5 | 686 | 102 | ||
BC106095.1 | - | 4 - 5 | 627 | 102 | ||
BC127163.1 | - | 4 - 5 | 573 | 102 | ||
BC152540.1 | - | 4 - 5 | 575 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
