Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800000598.mm.1 |
NM_033622.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800000598.mm.1 |
NM_033622.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800000598.mm.1 |
TC0800000074.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800000599.mm.1 |
NM_033622.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800000599.mm.1 |
NM_033622.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800000599.mm.1 |
TC0800000074.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800000597.mm.1 |
NM_033622.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800000597.mm.1 |
NM_033622.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800000597.mm.1 |
TC0800000074.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800000596.mm.1 |
NM_033622.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800000596.mm.1 |
NM_033622.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800000596.mm.1 |
TC0800000074.mm.1 |
MTA TC |