Assay Details
| Gene Symbol: Tnip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139064.2 | NP_620703.1 | 2 - 3 | 654 | 108 | |
| GenBank mRNA | AJ304865.1 | - | 2 - 3 | 654 | 108 | |
| AK160165.1 | - | 2 - 3 | 612 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tnip2
Genomic Map