Assay Details
| Gene Symbol: Ccnc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122982.2 | NP_001116454.1 | 6 - 7 | 577 | 167 | |
| NM_001290420.1 | NP_001277349.1 | 6 - 7 | 577 | 167 | ||
| NM_001290422.1 | NP_001277351.1 | 6 - 7 | 577 | 167 | ||
| NM_016746.4 | NP_058026.2 | 6 - 7 | 577 | 167 | ||
| GenBank mRNA | AK009615.1 | - | 6 - 7 | 522 | 167 | |
| AK014079.1 | - | 5 - 6 | 426 | 167 | ||
| AK037957.1 | - | 6 - 7 | 578 | 167 | ||
| AK135472.1 | - | 6 - 7 | 573 | 167 | ||
| AK153035.1 | - | 6 - 7 | 510 | 167 | ||
| BC003344.1 | - | 6 - 7 | 510 | 3257730 | 167 | |
| BC037689.1 | - | 6 - 7 | 505 | 167 | ||
| BC062376.1 | - | 6 - 7 | 492 | 167 | ||
| BC087544.1 | - | 6 - 7 | 509 | 167 | ||
| BC100396.1 | - | 6 - 7 | 551 | 167 | ||
| BC120649.1 | - | 6 - 7 | 504 | 40129572 | 167 | |
| BC120677.1 | - | 6 - 7 | 504 | 40129600 | 167 | |
| U62638.1 | - | 6 - 7 | 552 | 167 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccnc
Genomic Map