Assay Details
| Gene Symbol: Ccnc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122982.2 | NP_001116454.1 | 7 - 8 | 620 | 167 | |
| NM_001290420.1 | NP_001277349.1 | 7 - 8 | 620 | 167 | ||
| NM_001290422.1 | NP_001277351.1 | 7 - 8 | 620 | 167 | ||
| NM_016746.4 | NP_058026.2 | 7 - 8 | 620 | 167 | ||
| GenBank mRNA | AK009615.1 | - | 7 - 8 | 565 | 167 | |
| AK014079.1 | - | 6 - 7 | 469 | 167 | ||
| AK037957.1 | - | 7 - 8 | 621 | 167 | ||
| AK135472.1 | - | 7 - 8 | 616 | 167 | ||
| AK153035.1 | - | 7 - 8 | 553 | 167 | ||
| BC003344.1 | - | 7 - 8 | 553 | 3257730 | 167 | |
| BC037689.1 | - | 7 - 8 | 548 | 167 | ||
| BC062376.1 | - | 7 - 8 | 535 | 167 | ||
| BC087544.1 | - | 7 - 8 | 552 | 167 | ||
| BC100396.1 | - | 7 - 8 | 594 | 167 | ||
| BC120649.1 | - | 7 - 8 | 547 | 40129572 | 167 | |
| BC120677.1 | - | 7 - 8 | 547 | 40129600 | 167 | |
| U62638.1 | - | 7 - 8 | 595 | 167 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccnc
Genomic Map