Assay Details
Gene Symbol: Rad54b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039556.3 | NP_001034645.1 | 9 - 10 | 1565 | 127 | |
GenBank mRNA | BC057604.1 | - | 9 - 10 | 1513 | 6416213 | 127 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
