Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400001438.mm.1 |
NM_001122889.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400001438.mm.1 |
NM_001122889.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400001438.mm.1 |
NM_010141.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400001438.mm.1 |
TC0400000234.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400001441.mm.1 |
NM_001122889.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400001441.mm.1 |
NM_001122889.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400001441.mm.1 |
NM_010141.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400001441.mm.1 |
TC0400000234.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400001437.mm.1 |
NM_001122889.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400001437.mm.1 |
NM_001122889.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400001437.mm.1 |
NM_010141.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400001437.mm.1 |
TC0400000234.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400001439.mm.1 |
NM_001122889.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400001439.mm.1 |
NM_001122889.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400001439.mm.1 |
NM_010141.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400001439.mm.1 |
TC0400000234.mm.1 |
MTA TC |