Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400000762.mm.1 |
NM_001171801.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000762.mm.1 |
NM_001171801.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000762.mm.1 |
NM_173746.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000762.mm.1 |
NM_173746.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000762.mm.1 |
TC0400000114.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400000763.mm.1 |
NM_001171801.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000763.mm.1 |
NM_001171801.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000763.mm.1 |
NM_173746.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000763.mm.1 |
NM_173746.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000763.mm.1 |
TC0400000114.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400000764.mm.1 |
NM_001171801.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000764.mm.1 |
NM_001171801.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000764.mm.1 |
NM_173746.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000764.mm.1 |
NM_173746.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400000764.mm.1 |
TC0400000114.mm.1 |
MTA TC |