Assay Details
Gene Symbol: Triqk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171801.1 | NP_001165272.1 | 3 - 4 | 334 | 134 | |
GenBank mRNA | DQ351292.1 | - | 3 - 4 | 334 | 134 |

More Information
Additional Information:
This assay is in the 5'UTR region of the DQ351292.1, NM_001171801.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
