Assay Details
Gene Symbol: Kif24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK084967.1 | - | 3 - 4 | 961 | 137 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
