Assay Details
| Gene Symbol: Rbpj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080927.2 | NP_001074396.1 | 8 - 9 | 1105 | 151 | |
| NM_001080928.1 | NP_001074397.1 | 8 - 9 | 991 | 151 | ||
| NM_001277116.1 | NP_001264045.1 | 10 - 11 | 1187 | 151 | ||
| NM_009035.5 | NP_033061.3 | 8 - 9 | 1007 | 151 | ||
| GenBank mRNA | AK028363.1 | - | 10 - 11 | 1165 | 151 | |
| AK090006.1 | - | 8 - 9 | 892 | 151 | ||
| AK139087.1 | - | 8 - 9 | 892 | 151 | ||
| AK145187.1 | - | 8 - 9 | 899 | 151 | ||
| AK152746.1 | - | 8 - 9 | 1007 | 151 | ||
| AK153163.1 | - | 9 - 10 | 895 | 151 | ||
| BC035299.1 | - | 6 - 7 | 793 | 151 | ||
| BC051387.1 | - | 8 - 9 | 936 | 4460290 | 151 | |
| S63463.1 | - | 8 - 9 | 1008 | 151 | ||
| X17459.1 | - | 8 - 9 | 1003 | 151 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rbpj
Genomic Map