Assay Details
Gene Symbol: Tmco3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172282.2 | NP_758486.1 | 1 - 2 | 382 | 75 | |
GenBank mRNA | AK033499.1 | - | 1 - 2 | 143 | 75 | |
AK046065.1 | - | 1 - 2 | 172 | 75 | ||
AK049974.1 | - | 1 - 2 | 141 | 75 | ||
AK083321.1 | - | 1 - 2 | 206 | 75 | ||
AK088866.1 | - | 1 - 2 | 166 | 75 | ||
BC039955.1 | - | 1 - 2 | 128 | 4236297 | 75 | |
BC053487.1 | - | 1 - 2 | 157 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK033499.1, AK046065.1, AK049974.1, AK088866.1, BC039955.1, BC053487.1, NM_172282.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
