Assay Details
Gene Symbol: Tmco3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172282.2 | NP_758486.1 | 6 - 7 | 1489 | 75 | |
GenBank mRNA | AK033499.1 | - | 6 - 7 | 1250 | 75 | |
AK046065.1 | - | 6 - 7 | 1279 | 75 | ||
AK049974.1 | - | 6 - 7 | 1248 | 75 | ||
AK088866.1 | - | 6 - 7 | 1273 | 75 | ||
AK148252.1 | - | 6 - 7 | 1292 | 75 | ||
BC039955.1 | - | 6 - 7 | 1235 | 4236297 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
