Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500019714.mm.1 |
TC0500002169.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500019714.mm.1 |
NM_016703.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500019710.mm.1 |
TC0500002169.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500019710.mm.1 |
NM_016703.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500019711.mm.1 |
TC0500002169.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500019711.mm.1 |
NM_016703.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500019719.mm.1 |
TC0500002169.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500019719.mm.1 |
NM_016703.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500019715.mm.1 |
TC0500002169.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500019715.mm.1 |
NM_016703.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500019720.mm.1 |
TC0500002169.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500019720.mm.1 |
NM_016703.3 |
Refseq |