Assay Details
Gene Symbol: Zbtb49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC028988.1 | - | 3 - 4 | 1339 | 5376301 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
