Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500005826.mm.1 |
TC0500000657.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500005826.mm.1 |
NM_001018019.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500005826.mm.1 |
NM_001018019.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500005827.mm.1 |
TC0500000657.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500005827.mm.1 |
NM_001018019.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500005827.mm.1 |
NM_001018019.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500005828.mm.1 |
TC0500000657.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500005828.mm.1 |
NM_001018019.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500005828.mm.1 |
NM_001018019.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500005829.mm.1 |
TC0500000657.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500005829.mm.1 |
NM_001018019.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500005829.mm.1 |
NM_001018019.2 |
Refseq |