Assay Details
| Gene Symbol: Mcph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173189.2 | NP_775281.2 | 2 - 3 | 556 | 78 | |
| GenBank mRNA | AK042026.1 | - | 2 - 3 | 565 | 78 | |
| AK083572.1 | - | 2 - 3 | 599 | 78 | ||
| AK138858.1 | - | 2 - 3 | 575 | 78 | ||
| AK147997.1 | - | 2 - 3 | 148 | 78 | ||
| AK148021.1 | - | 2 - 3 | 154 | 78 | ||
| AY070216.1 | - | 2 - 3 | 136 | 78 | ||
| BC056924.1 | - | 2 - 3 | 556 | 5697978 | 78 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK042026.1 transcripts.
Set Membership: |
|

Mcph1
Genomic Map