Assay Details
| Gene Symbol: Mcph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173189.2 | NP_775281.2 | 5 - 6 | 870 | 68 | |
| GenBank mRNA | AK083572.1 | - | 5 - 6 | 913 | 68 | |
| AK147997.1 | - | 5 - 6 | 462 | 68 | ||
| AK148021.1 | - | 5 - 6 | 468 | 68 | ||
| AY070216.1 | - | 5 - 6 | 450 | 68 | ||
| BC056924.1 | - | 5 - 6 | 870 | 5697978 | 68 | |
More Information
Set Membership: |
|

Mcph1
Genomic Map