Assay Details
Gene Symbol: Mx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK076430.1 | - | 11 - 12 | 1296 | 122 | |
M21039.1 | - | 8 - 9 | 1120 | 122 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK076430.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
