Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600016610.mm.1 |
NR_003520.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600016610.mm.1 |
NR_003520.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600016610.mm.1 |
TC1600002148.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600016608.mm.1 |
NR_003520.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600016608.mm.1 |
NR_003520.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600016608.mm.1 |
TC1600002148.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600016609.mm.1 |
NR_003520.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600016609.mm.1 |
NR_003520.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600016609.mm.1 |
TC1600002148.mm.1 |
MTA TC |