Assay Details
Gene Symbol: Lrrc71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028971.1 | NP_083247.1 | 12 - 13 | 1489 | 68 | |
GenBank mRNA | AK016993.1 | - | 12 - 13 | 1417 | 68 | |
AK029938.1 | - | 12 - 13 | 1457 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
