Assay Details
Gene Symbol: Lrrc71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028971.1 | NP_083247.1 | 4 - 5 | 669 | 98 | |
GenBank mRNA | AK016993.1 | - | 4 - 5 | 597 | 98 | |
AK029938.1 | - | 4 - 5 | 637 | 98 | ||
BB614807.1 | - | 4 - 5 | 669 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
