Assay Details
| Gene Symbol: Creb3l4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB182651.1 | - | 1 - 2 | 202 | 106 | |
| AK019928.1 | - | 1 - 2 | 202 | 106 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB182651.1, AK019928.1 transcripts.
Set Membership: |
|

Creb3l4
Genomic Map