Assay Details
Gene Symbol: Gfm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138591.2 | NP_613057.2 | 13 - 14 | 1723 | 95 | |
GenBank mRNA | AF315511.1 | - | 13 - 14 | 1690 | 95 | |
BC031772.1 | - | 13 - 14 | 1690 | 4239980 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
