Assay Details
| Gene Symbol: Shox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013665.1 | NP_038693.1 | 2 - 3 | 768 | 138 | |
| GenBank mRNA | AA509933.1 | - | 2 - 3 | 252 | 138 | |
| AK032007.1 | - | 2 - 3 | 315 | 138 | ||
| AK045937.1 | - | 1 - 2 | 210 | 138 | ||
| AK145063.1 | - | 2 - 3 | 264 | 138 | ||
| U66918.1 | - | 2 - 3 | 768 | 138 | ||
| U67055.1 | - | 2 - 3 | 283 | 138 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Shox2
Genomic Map