Assay Details
Gene Symbol: Shox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013665.1 | NP_038693.1 | 3 - 4 | 825 | 96 | |
GenBank mRNA | AA509933.1 | - | 3 - 4 | 309 | 96 | |
AK032007.1 | - | 3 - 4 | 372 | 96 | ||
AK045937.1 | - | 2 - 3 | 267 | 96 | ||
AK145063.1 | - | 3 - 4 | 321 | 96 | ||
U66918.1 | - | 3 - 4 | 825 | 96 | ||
U67055.1 | - | 3 - 4 | 340 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
