Assay Details
| Gene Symbol: Gatb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144896.4 | NP_659145.1 | 10 - 11 | 1341 | 81 | |
| GenBank mRNA | AK028193.1 | - | 10 - 11 | 1340 | 81 | |
| AK034702.1 | - | 10 - 11 | 1342 | 81 | ||
| AK053978.1 | - | 10 - 11 | 1344 | 81 | ||
| AK081800.1 | - | 10 - 11 | 1345 | 81 | ||
| AK134831.1 | - | 10 - 11 | 1203 | 81 | ||
| AK166231.1 | - | 10 - 11 | 1352 | 81 | ||
| BC005709.1 | - | 10 - 11 | 1343 | 3157973 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gatb
Genomic Map