Assay Details
| Gene Symbol: Gatb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144896.4 | NP_659145.1 | 7 - 8 | 974 | 97 | |
| GenBank mRNA | AK028193.1 | - | 7 - 8 | 973 | 97 | |
| AK034702.1 | - | 7 - 8 | 975 | 97 | ||
| AK053978.1 | - | 7 - 8 | 977 | 97 | ||
| AK081800.1 | - | 7 - 8 | 978 | 97 | ||
| AK134831.1 | - | 7 - 8 | 836 | 97 | ||
| AK166231.1 | - | 7 - 8 | 985 | 97 | ||
| BC005709.1 | - | 7 - 8 | 976 | 3157973 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gatb
Genomic Map