Assay Details
Gene Symbol: Pmf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025928.3 | NP_080204.1 | 4 - 5 | 562 | 69 | |
GenBank mRNA | AF348509.1 | - | 2 - 3 | 365 | 69 | |
AF348510.1 | - | 4 - 5 | 572 | 69 | ||
AK011174.1 | - | 4 - 5 | 564 | 69 | ||
AK012113.1 | - | 4 - 5 | 563 | 69 | ||
AK013787.1 | - | 4 - 5 | 565 | 69 | ||
AK078308.1 | - | 4 - 5 | 582 | 69 | ||
AK150603.1 | - | 4 - 5 | 579 | 69 | ||
AK168200.1 | - | 4 - 5 | 563 | 69 | ||
AK194541.1 | - | 1 - 2 | 59 | 69 | ||
BC034333.1 | - | 4 - 5 | 540 | 69 | ||
BC037139.1 | - | 4 - 5 | 571 | 4952550 | 69 | |
BC049223.1 | - | 4 - 5 | 561 | 3257320 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
cell cycle
chromosome segregation
mitotic nuclear division
cell division
regulation of nucleic acid-templated transcription
transcription coactivator activity
protein binding
leucine zipper domain binding
transcription, DNA-templated
regulation of transcription, DNA-templated
cell cycle
chromosome segregation
mitotic nuclear division
cell division
regulation of nucleic acid-templated transcription
transcription coactivator activity
protein binding
leucine zipper domain binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22313617 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22313619 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22313622 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22313620 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22313618 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P438859 | AK011174.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1966528 | BC034333.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1966528 | BC037139.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1966528 | BC049223.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1966528 | AK011174.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1966528 | AK012113.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 953957 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 478908 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 926602 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 75515 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 778456 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 24307 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 564742 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 173719 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 103858 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P102496 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P120961 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P438859 | AK011174.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1966528 | BC034333.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1966528 | BC037139.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1966528 | BC049223.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1966528 | AK011174.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1966528 | AK012113.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313622 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313619 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313614 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313610 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313616 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313621 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313623 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313615 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313607 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313609 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313611 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04980806 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P120961 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00694774 | - | - |
Whole Mouse Genome G4122A | A_51_P438865 | BC034333.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P438865 | BC037139.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P438865 | BC049223.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P438865 | AK011174.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P438865 | AK012113.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123019 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P120961 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02368617 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02368607 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22313618 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22313620 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22313622 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22313619 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22313617 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313549 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313552 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313539 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313521 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313547 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313535 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313602 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313564 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313572 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313554 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313530 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313517 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313524 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313576 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313611 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313548 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313531 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313546 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313615 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313536 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313525 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313507 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313565 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313593 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313570 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313516 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313515 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313675 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313568 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313589 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313545 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313542 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313567 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313533 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313555 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313551 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313621 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313614 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313522 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313603 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313604 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313582 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313599 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313508 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313591 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313679 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313610 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313594 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313619 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313559 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313511 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313509 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02566831 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03084227 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02983286 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02916118 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22313621 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22313589 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22313582 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22313603 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03039431 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22313599 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02595535 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22313622 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02778967 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02728597 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22313611 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03084230 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22313623 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22313615 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22313585 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22313586 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22313591 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22313619 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22313607 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02949287 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22313597 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22313610 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10499367 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10499371 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10499368 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10499370 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10499369 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1438173_PM_x_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1416707_PM_a_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10499366 | - | - |
Mouse Genome 430 2.0 Array | 1438173_PM_x_at | - | - |
Mouse Genome 430 2.0 Array | 1416707_a_at | - | - |
Mouse Genome 430 2.0 Array | 1416707_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1438173_x_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67037_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67037_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0300017508.mm.1 | NM_025928.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300017508.mm.1 | TC0300002310.mm.1 | MTA TC |
Mouse Expression Set 430 | 1416707_PM_a_at | - | - |
Mouse Expression Set 430 | 1438173_x_at | - | - |
Mouse Expression Set 430 | 1416707_a_at | - | - |
Mouse Expression Set 430 | 1438173_PM_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1438173_PM_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1438173_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1416707_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1416707_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1416707_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1416707_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1438173_x_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1438173_PM_x_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 5130273 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 5130273 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005130273 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2711410 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2711410 | - | - |
mouse-6 v1.1 (Illumina) | scl067037.1_35-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5130273 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl067037.1_35-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5130273 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2711410 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2711410 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl067037.1_35-S | - | - |
