Assay Details
| Gene Symbol: Stard13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163493.1 | NP_001156965.1 | 11 - 12 | 2987 | 70 | |
| NM_146258.2 | NP_666370.3 | 11 - 12 | 2930 | 70 | ||
| GenBank mRNA | AK050259.1 | - | 11 - 12 | 2988 | 70 | |
| AK136202.1 | - | 7 - 8 | 1513 | 70 | ||
| BC027830.1 | - | 11 - 12 | 2942 | 5320966 | 70 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Stard13
Genomic Map