Assay Details
Gene Symbol: 1600002D24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040485.1 | 3 - 4 | 478 | 67 | ||
GenBank mRNA | AK037512.1 | - | 3 - 4 | 478 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P549892 | AK005393.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P549892 | AK005393.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 181260 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 308775 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 198572 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 185701 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 508449 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 379035 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 418644 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 658430 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 766635 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 155691 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 887775 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 923015 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 51478 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 793569 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 349812 | AK034419.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 34560 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 495750 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 876880 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 260375 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 460257 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 576280 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 921578 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 77747 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 96507 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 200511 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 39851 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 131702 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 275393 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 862111 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 430745 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 810012 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 11411 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 44074 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 127823 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 137916 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 85347 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P549892 | AK005393.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P549892 | AK005393.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03264098 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08694064 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03264122 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08694147 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08694045 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03264178 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03264158 | - | - |
Whole Mouse Genome G4122A | A_52_P549892 | AK005393.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P549892 | AK005393.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116358 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08694045 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P03264122 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P03264158 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989781 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989959 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12754689 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989759 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989906 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989913 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989793 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12762454 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12908352 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12908365 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30990098 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30990105 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989873 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12514539 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989971 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989975 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12716763 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989787 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989765 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989877 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989794 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12559859 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989790 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989871 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989763 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12908355 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989779 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989882 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989896 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30990097 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989961 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989872 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989788 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30990094 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989748 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989789 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989696 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989699 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989757 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30990092 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989799 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989903 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989868 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989746 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30990101 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989782 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12826162 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989964 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989698 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989745 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989800 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12908149 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989760 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989978 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989932 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12769221 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989749 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989914 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12812619 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12856117 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989933 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989966 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989795 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989927 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989810 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989797 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989974 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989750 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989879 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989929 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30990093 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989909 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12612961 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12716765 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12606969 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30990099 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30990103 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12596269 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989924 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989801 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12564964 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989982 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989694 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12504369 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989808 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989956 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12503031 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30990104 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989803 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12606980 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989905 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12762453 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989968 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989953 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989951 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12655053 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30990029 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989791 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989902 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12908143 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12826157 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12673449 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989904 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989907 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30990035 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12504372 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989972 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12908148 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30989970 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12856112 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Clariom S Array | PSR1600016379.mm.1 | NR_040484.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600016379.mm.1 | NR_040484.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600016379.mm.1 | NR_040485.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600016379.mm.1 | NR_040485.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600016379.mm.1 | TC1600002134.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1600016381.mm.1 | NR_040485.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600016381.mm.1 | NR_040485.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600016381.mm.1 | TC1600002134.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1600016376.mm.1 | NR_040485.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600016376.mm.1 | NR_040485.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600016376.mm.1 | TC1600002134.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1600016380.mm.1 | NR_040485.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600016380.mm.1 | NR_040485.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1600016380.mm.1 | TC1600002134.mm.1 | MTA TC |
Murine Genome U74v2 Set | 169674_at | - | - |
Murine Genome U74v2 Set | 169674_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 645691 | - | - |
