Assay Details
| Gene Symbol: Fam219aos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045726.1 | 1 - 2 | 242 | 107 | ||
| GenBank mRNA | AK006904.1 | - | 1 - 2 | 243 | 107 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 1110017D15 gene | 1110017D15Rik |
Cbe1, Smrp1 |
-104.0 | Anti-sense | Chr. 4 - 41505005 - 41517333 on NCBI Build GRCm38 |
| Downstream | predicted gene 12404 | Gm12404 | OTTMUSG00000006660 | 2.2kb | Sense | Chr. 4 - 41520405 - 41522396 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK006904.1 transcripts.
Set Membership: |
|

Fam219aos
Genomic Map