Assay Details
| Gene Symbol: Gm31343 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK042586.1 | - | 3 - 4 | 408 | 155 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
solute carrier family 10, member 2 |
Slc10a2 |
ASBT, ISBT |
-3.3mb | Anti-sense | Chr. 8 - 5085623 - 5105232 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 31135 |
Gm31135 | 54.1kb | Anti-sense | Chr. 8 - 8436606 - 8484728 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm31343
Genomic Map