Assay Details
Gene Symbol: Gm31463 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK053231.1 | - | 3 - 4 | 753 | 57 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
predicted gene, 31135 |
Gm31135 | -93.8kb | Anti-sense | Chr. 8 - 8436606 - 8484728 on NCBI Build GRCm38 | |
Downstream | ephrin B2 | Efnb2 |
ELF-2, Epl5, Eplg5, Htk-L, LERK-5, Lerk5, NLERK-1 |
36.9kb | Anti-sense | Chr. 8 - 8617235 - 8660773 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
