Assay Details
| Gene Symbol: Mapk8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016700.4 | NP_057909.1 | 3 - 4 | 450 | 93 | |
| GenBank mRNA | AB219977.1 | - | 2 - 3 | 259 | 93 | |
| AK030767.1 | - | 3 - 4 | 452 | 93 | ||
| AK032486.1 | - | 3 - 4 | 448 | 93 | ||
| AK043102.1 | - | 3 - 4 | 450 | 93 | ||
| AK139498.1 | - | 3 - 4 | 438 | 93 | ||
| BB619635.1 | - | 3 - 4 | 452 | 93 | ||
| BC053027.1 | - | 3 - 4 | 420 | 5694771 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mapk8
Genomic Map