Assay Details
| Gene Symbol: Slc7a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301424.1 | NP_001288353.1 | 5 - 6 | 850 | 70 | |
| NM_007513.4 | NP_031539.3 | 5 - 6 | 966 | 70 | ||
| GenBank mRNA | AK139342.1 | - | 5 - 6 | 942 | 70 | |
| AK147991.1 | - | 5 - 6 | 875 | 70 | ||
| AK163202.1 | - | 5 - 6 | 850 | 70 | ||
| BC068234.1 | - | 5 - 6 | 857 | 69 | ||
| BC145779.1 | - | 4 - 5 | 771 | 40131082 | 70 | |
| BC145781.1 | - | 4 - 5 | 771 | 40131084 | 70 | |
| M26687.1 | - | 5 - 6 | 884 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc7a1
Genomic Map